missing translation for 'onlineSavingsMsg'
Learn More

Disuccinimidyl glutarate, 97%

Catalog No. p-7039083
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58208MC 100 mg
AAH58208ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH58208MC Supplier Thermo Scientific Chemicals Supplier No. H58208MC

CAS: 79642-50-5 | C13H14N2O8 | 326.26 g/mol

Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 79642-50-5
Molecular Formula C13H14N2O8
Molecular Weight (g/mol) 326.26
MDL Number MFCD00153597
InChI Key LNQHREYHFRFJAU-UHFFFAOYSA-N
Synonym disuccinimidyl glutarate, di n-succinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl pentanedioate, pentanedioic acid,1,5-bis 2,5-dioxo-1-pyrrolidinyl ester, dsg crosslinker, glutaric acid disuccinimidyl ester, di n-hydroxysuccinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl glutarate, di n-succinimidyl glutarate chn
PubChem CID 4432628
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) pentanedioate
SMILES O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O

Specifications

Quantity 100 mg
Beilstein 8575145
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Formula Weight 326.26
Percent Purity 97%
Chemical Name or Material Disuccinimidyl glutarate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only