accessibility menu, dialog, popup

UserName

CAS RN 79642-50-5

IUPAC Name: bis(2,5-dioxopyrrolidin-1-yl) pentanedioate
Synonyms: bis(2,5-dioxopyrrolidin-1-yl) pentanedioate
Molecular Weight (g/mol): 326.26
Molecular Formula: C13H14N2O8
InChi Key: LNQHREYHFRFJAU-UHFFFAOYSA-N
SMILES: O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results