missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Disposable Inoculation Loops

Save time and eliminate cross-contamination

$148.54 - $308.06

Specifications

Length (English) 9 in.
Length (Metric) 22.7 cm
Material Polystyrene
Sterility Sterile
Disposable Single-use
View More Specs

Products 3
Catalog Number Mfr. No. Volume Color Quantity Price Quantity  
Catalog Number Mfr. No. Volume Color Quantity Price Quantity  
03-391-561
View Documents
Cole-Parmer™ Essentials
0623104
10 μL Yellow 40
Case of 40 Each for $308.06
Only null left
 
03-391-565
View Documents
Cole-Parmer™ Essentials
0623108
10 μL Yellow 250
Case of 250 Each for $148.54
Only null left
 
03-391-564
View Documents
Cole-Parmer™ Essentials
0623107
1 μL Blue 250
Case of 250 Each for $148.69
Only null left
 
Description

Description

  • Color coded
  • Sterile

Smooth and flexible loops are constructed of high-impact polystyrene. Loops ensure uniform streaking without damaging your gel surface. Color coding identifies inoculation volume easily. Packed sterile in tamperproof resealable bags. Each case is supplied with a quality certificate confirming sterility and accuracy.

Specifications

Specifications

9 in.
Polystyrene
Single-use
22.7 cm
Sterile
Inoculating Loop
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'