missing translation for 'onlineSavingsMsg'
Learn More

Disodium Terephthalate 99.0+%, TCI America™

Catalog No. p-7136604
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T109725G 25 g
T1097100G 100 g
T1097500G 500 g
3 options

Catalog No. T109725G

Supplier: TCI America T109725G

Only null left

Chemical Identifiers

CAS 10028-70-3
Molecular Formula C8H4Na2O4
Molecular Weight (g/mol) 210.096
MDL Number MFCD00013137
InChI Key VIQSRHWJEKERKR-UHFFFAOYSA-L
Synonym disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:?
PubChem CID 82305
IUPAC Name disodium;terephthalate
SMILES C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 210.10
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Terephthalate
EINECSNumber (3)-0073&(3)-1272
RTECSNumber WZ1400000
TSCA No