accessibility menu, dialog, popup

UserName

CAS RN 10028-70-3

IUPAC Name: disodium benzene-1,4-dicarboxylate
Synonyms: disodium terephthalate
Molecular Weight (g/mol): 210.1
Molecular Formula: C8H4Na2O4
InChi Key: VIQSRHWJEKERKR-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=C(C=C1)C([O-])=O
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)