missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ HS C18 Prep Guard Cartridge, 10 μm Particle Size

Catalog No. 111009163
Change view
Click to view available options
Diameter: 21.2 mm; Length: 1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111009163 Diameter: 21.2 mm; Length: 1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009163 Supplier MilliporeSigma™ Supelco™ Supplier No. 581394U

Specifications

Quantity 1/Pk.
Particle Size 10 μm
Length (Metric) 1 cm
Pore Size 120 Å
Type Guard Cartridge
Diameter (Metric) Inner 21.2 mm
Phase Reversed Phase
For Use With (Application) HPLC
Product Line Discovery

Discovery is a registered trademark of Sigma-Aldrich Co. LLC