missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size

Catalog No. 111008673
Change view
Click to view available options
Length (Metric):
2 cm
5 cm
10 cm
12.5 cm
15 cm
25 cm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
10 mm
21.2 mm
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111008673 2 cm 2.1 mm
111008465 5 cm 2.1 mm
111007417 10 cm 2.1 mm
111007306 12.5 cm 2.1 mm
111007292 15 cm 2.1 mm
111007251 25 cm 2.1 mm
111007204 15 cm 3 mm
111007013 25 cm 3 mm
111007418 10 cm 4 mm
111007307 12.5 cm 4 mm
111007205 15 cm 4 mm
111007014 25 cm 4 mm
111020843 5 cm 4.6 mm
111007419 10 cm 4.6 mm
111016245 12.5 cm 4.6 mm
111007012 25 cm 4.6 mm
111016199 25 cm 10 mm
111005710 25 cm 21.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 111008673 Supplier MilliporeSigma™ Supelco™ Supplier No. 577507U

Use Discovery™ C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.

  • Excellent reproducibility
  • Exceptional peak shape for basic and acidic analytes
  • Stable, low-bleed LC-MS separations
  • Separation of peptides and small proteins
  • Lower hydrophobicity than many comparable C18 columns, providing faster analysis

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 2 cm
Particle Size 5 μm
pH 2 to 8
Pore Size 180 Å
Surface Area 200 m2/g
Stationary Phase C18 (Octadecyl)
Product Line Discovery
Matrix C18 (Octadecyl) Phase/Silica Gel, High Purity, Spherical Particle Platform
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Product Type HPLC Column
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC