missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size
Description
- Excellent reproducibility
- Exceptional peak shape for basic and acidic analytes
- Stable, low-bleed LC-MS separations
- Separation of peptides and small proteins
- Lower hydrophobicity than many comparable C18 columns, providing faster analysis
Specifications
Specifications
| Column Format | HPLC, LC/MS Column |
| Diameter (Metric) | 4 mm |
| Length (Metric) | 15 cm |
| Max. Pressure | 400 bar(5801 psi) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 180 Å |
| Surface Area | 200 m2/g |
| Temperature | ≤70°C |
| USP Type | USP L1 |
| Show More |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC