missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size
Use Discovery™ C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.
Supplier: MilliporeSigma™ Supelco™ 569226U
Description
- Excellent reproducibility
- Exceptional peak shape for basic and acidic analytes
- Stable, low-bleed LC-MS separations
- Separation of peptides and small proteins
- Lower hydrophobicity than many comparable C18 columns, providing faster analysis
Specifications
| HPLC Column | |
| 25 cm | |
| 5 μm | |
| 180 Å | |
| ≤70°C | |
| C18 (Octadecyl) | |
| C18 (Octadecyl) Phase/Silica Gel, High Purity, Spherical Particle Platform | |
| 12% | |
| Yes | |
| Stainless Steel | |
| HPLC |
| 21.2 mm | |
| 400 bar(5801 psi) | |
| 2 to 8 | |
| 200 m2/g | |
| USP L1 | |
| Discovery | |
| Silica gel, high purity, spherical particle platform fully porous particle | |
| Reversed Phase | |
| Reversed Phase | |
| HPLC Column |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC