missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C8 HPLC Column, 10 μm Particle Size

Catalog No. 111005770
Change view
Click to view available options
Length (Metric):
25 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111005770 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005770 Supplier MilliporeSigma™ Supelco™ Supplier No. 567225U

Discovery™ BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery™ BIO Wide Pore C5 and the Discovery™ BIO Wide Pore C18.

Discovery™ BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery™ BIO Wide Pore C5 and the Discovery™ BIO Wide Pore C18. The difference in hydrophobicity gives it unique selectivity relative to these other phases. It is ideal for peptide mapping because it provides complementary information compared to a C18 separation. Because of its intermediate hydrophobicity, we also recommend it for method development or scouting work. As with all Discovery™ BIO Wide Pore phases, the C8 phase gives efficient, symmetrical peaks, exceptional stability, long column lifetime, and LC-MS compatibility.

Specifications

Diameter (Metric) 21.2 mm
Length (Metric) 25 cm
Particle Size 10 μm
Pore Size 300 Å
Surface Area 100 m2/g
Stationary Phase C8 (Octyl)
Product Line Discovery
Matrix C8 (Octyl) Phase/silica particle platform
Endcapped Yes
Phase Reversed Phase
Technique Wide Pore HPLC
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC