missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C5 HPLC Column, 3 μm Particle Size
Discovery™ BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC.
Supplier: MilliporeSigma™ Supelco™ 567230U
Description
Discovery™ BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC. Long-chain phases, like C8 or C18, are often too hydrophobic for proteins and can cause excessively long retention time or even irreversible binding to the column. For this reason short-chain phases, typically C3 or C4, are often used for RP-HPLC of proteins. However, these short-chain phases are susceptible to hydrolysis, resulting in short column lifetime, especially at low pH. The Discovery™ BIO Wide Pore C5 gives elution order similar to a conventional C4, yet has enhanced pH stability for longer column lifetime. Generally, higher efficiency separations are achievable on the Discovery™ BIO Wide Pore C5 because of the improvements we have made to the silica and bonded-phase chemistry.Specifications
| 4.6 mm | |
| 3 μm | |
| 100 m2/g | |
| Discovery | |
| Yes | |
| Wide Pore HPLC |
| 10 cm | |
| 300 Å | |
| C5 (pentyl) | |
| C5 (pentyl) Phase/Silica Particle Platform | |
| Reversed Phase |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC