missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C5 HPLC Column, 10 μm Particle Size

Catalog No. p-7189974
Change view
Click to view available options
Length (Metric):
15 cm
25 cm
Diameter (Metric):
4.6 mm
10 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111007403 25 cm 4.6 mm
111006661 15 cm 10 mm
111006415 25 cm 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111007403 Supplier MilliporeSigma™ Supelco™ Supplier No. 567232U
Only null left

Discovery™ BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC.

Discovery™ BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC. Long-chain phases, like C8 or C18, are often too hydrophobic for proteins and can cause excessively long retention time or even irreversible binding to the column. For this reason short-chain phases, typically C3 or C4, are often used for RP-HPLC of proteins. However, these short-chain phases are susceptible to hydrolysis, resulting in short column lifetime, especially at low pH. The Discovery™ BIO Wide Pore C5 gives elution order similar to a conventional C4, yet has enhanced pH stability for longer column lifetime. Generally, higher efficiency separations are achievable on the Discovery™ BIO Wide Pore C5 because of the improvements we have made to the silica and bonded-phase chemistry.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 25 cm
Particle Size 10 μm
Pore Size 300 Å
Surface Area 100 m2/g
Stationary Phase C5 (pentyl)
Product Line Discovery
Matrix C5 (pentyl) Phase/Silica Particle Platform
Endcapped Yes
Phase Reversed Phase
Technique Wide Pore HPLC
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC