missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C18 HPLC Column, 3 μm Particle Size
The Discovery™ BIO Wide Pore C18 column has the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.
Supplier: MilliporeSigma™ Supelco™ 567202U
Description
Peptide maps generated by RP-HPLC provide valuable information about protein structure, stability, and purity. To be effective, the RP-HPLC column must be able to resolve a high percentage of the peptides in the sample. The more peptides, the better the information. Discovery™ BIO Wide Pore C18 gives unsurpassed RP-HPLC resolution of peptide maps from enzymatic digests. The improvements in silica and bonded-phase chemistry that have been incorporated into the Discovery™ BIO Wide Pore line improve resolution by increasing efficiency and reducing the peak tailing. An added benefit to this is the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.Specifications
| 2.1 mm | |
| 3 μm | |
| 100 m2/g | |
| Discovery | |
| Yes | |
| Wide Pore HPLC |
| 15 cm | |
| 300 Å | |
| C18 (octadecyl) | |
| C18 (octadecyl) Phase/silica particle platform | |
| Reversed Phase |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC