missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C18 HPLC Column, 10 μm Particle Size

Catalog No. p-7189960
Change view
missing translation for 'orderingAttributeHoverText'
Length (Metric):
15 cm
25 cm
5 cm
Diameter (Metric):
10 mm
21.2 mm
Catalog No. Length (Metric) Diameter (Metric)
111003247 5 cm 10 mm
111003065 15 cm 10 mm
111002611 25 cm 10 mm
111002880 15 cm 21.2 mm
111002824 25 cm 21.2 mm
5 options

Catalog No. 111003247

Supplier: MilliporeSigma™ Supelco™ 567207U

Only null left

The Discovery™ BIO Wide Pore C18 column has the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.

Peptide maps generated by RP-HPLC provide valuable information about protein structure, stability, and purity. To be effective, the RP-HPLC column must be able to resolve a high percentage of the peptides in the sample. The more peptides, the better the information. Discovery™ BIO Wide Pore C18 gives unsurpassed RP-HPLC resolution of peptide maps from enzymatic digests. The improvements in silica and bonded-phase chemistry that have been incorporated into the Discovery™ BIO Wide Pore line improve resolution by increasing efficiency and reducing the peak tailing. An added benefit to this is the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.

Specifications

Diameter (Metric) 10 mm
Length (Metric) 5 cm
Particle Size 10 μm
Pore Size 300 Å
Surface Area 100 m2/g
Stationary Phase C18 (octadecyl)
Product Line Discovery
Matrix C18 (octadecyl) Phase/silica particle platform
Endcapped Yes
Phase Reversed Phase
Technique Wide Pore HPLC
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC