missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium (R)-1,1'-Binaphthyl-2,2'-disulfonate 98.0+%, TCI America™

Catalog No. D4445100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4445100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4445100MG Supplier TCI America Supplier No. D4445100MG
Only null left

Chemical Identifiers

CAS 1092934-19-4
Molecular Formula C20H12K2O6S2
Molecular Weight (g/mol) 490.627
InChI Key ALWKFXOUXLSOSQ-UHFFFAOYSA-L
Synonym (R)-BINSA Dipotassium Salt, (R)-1,1′C-Binaphthyl-2,2′C-disulfonic Acid Dipotassium Salt
PubChem CID 87548207
IUPAC Name dipotassium;1-(2-sulfonatonaphthalen-1-yl)naphthalene-2-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)S(=O)(=O)[O-])S(=O)(=O)[O-].[K+].[K+]

Specifications

Color White
Quantity 100 mg
Formula Weight 490.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dipotassium (R)-1,1′-Binaphthyl-2,2′-disulfonate