missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium 2-Naphthol-6,8-disulfonate Hydrate 98.0+%, TCI America™

Catalog No. N003025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N003025G Supplier TCI America Supplier No. N003025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 842-18-2
Molecular Formula C10H6K2O7S2
Molecular Weight (g/mol) 380.47
MDL Number MFCD00066361
InChI Key LKDMVWRBMGEEGR-UHFFFAOYSA-L
Synonym dipotassium 2-naphthol-6,8-disulfonate, dipotassium 7-hydroxynaphthalene-1,3-disulphonate, unii-i4s7vvu23a, 1,3-naphthalenedisulfonic acid, 7-hydroxy-, dipotassium salt, 2-naphthol-6,8-disulfonic acid dipotassium salt, dipotassium 7-hydroxynaphthalene-1,3-disulfonate, i4s7vvu23a, dipotassium 7-hydroxy-1,3-naphthalenedisulfonate, 7-hydroxy-1,3-naphthalenedisulfonic acid dipotassium salt, g acid dipotassium salt
PubChem CID 70050
IUPAC Name dipotassium 7-hydroxynaphthalene-1,3-disulfonate
SMILES [K+].[K+].OC1=CC=C2C=C(C=C(C2=C1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Gray-White
Quantity 25 g
Formula Weight 380.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dipotassium 2-Naphthol-6,8-disulfonate Hydrate
EINECSNumber (4)-0498
TSCA Yes