accessibility menu, dialog, popup

UserName

KKHOSOOOSOOO

CAS RN 842-18-2

IUPAC Name: dipotassium 7-hydroxynaphthalene-1,3-disulfonate
Synonyms: dipotassium 7-hydroxynaphthalene-1,3-disulfonate
Molecular Weight (g/mol): 380.47
Molecular Formula: C10H6K2O7S2
InChi Key: LKDMVWRBMGEEGR-UHFFFAOYSA-L
SMILES: [K+].[K+].OC1=CC=C2C=C(C=C(C2=C1)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results