missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane 99.0+%, TCI America™

Catalog No. D0896500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0896500G 500 g
D089625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0896500G Supplier TCI America Supplier No. D0896500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 500 g
UN Number 3077
Formula Weight 168.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Diphenylmethane
EINECSNumber (4)-0085
RTECSNumber DA8990000
TSCA Yes