accessibility menu, dialog, popup

UserName

CAS RN 101-81-5

IUPAC Name: benzylbenzene
Synonyms: diphenylmethane 1,1'-dimethylenebis(benzene) ditan ditane diphenyl methane
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: CZZYITDELCSZES-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)