missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane, 99%

CAS: 101-81-5 | C13H12 | 168.24 g/mol

$192.30 - $192.30

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117350010
SDS View Documents
Thermo Scientific Chemicals
117350010
1 kg Glass bottle
Each for $192.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-81-5
168.24
CZZYITDELCSZES-UHFFFAOYSA-N
7580
benzylbenzene
C13H12
MFCD00004781
diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
CHEBI:38884
C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

101-81-5
Colorless to Yellow
265.0°C
Authentic
Glass bottle
1.5760 to 1.5780
MFCD00004781
05, II, 498
15, 3362
Solubility in water: practically insoluble. Other solubilities: insoluble in liquid ammonia, freely soluble in alcohol, ether, chloroform,, hexane and benzene
C(C1=CC=CC=C1)C1=CC=CC=C1
168.24
CHEBI:38884
99%
Diphenylmethane, 99%
24.5°C to 25.0°C
1.0000g/mL
127°C
98.5% min. (GC)
C13H12
(C6H5)2CH2
1 kg
1
diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
CZZYITDELCSZES-UHFFFAOYSA-N
benzylbenzene
7580
168.24
Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 202-978-6

RTECSNumber : DA4976500

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only