missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane, 99+%

Catalog No. p-7025646
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2125622 100 g
AAB2125636 500 g
AAB212560E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2125622 Supplier Thermo Scientific Chemicals Supplier No. B2125622
Only null left

CAS: 101-81-5 | C13H12 | 168.24 g/mol

Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

Solubility
Soluble in ethanol, ether, benzene and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 22°C to 26°C
Density 1.01
Boiling Point 262°C to 264°C
Flash Point 130°C (266°F)
Refractive Index 1.577
Quantity 100 g
UN Number UN3077
Beilstein 1904982
Merck Index 14,3328
Solubility Information Soluble in ethanol,ether,benzene and chloroform.
Formula Weight 168.24
Percent Purity ≥99%
Chemical Name or Material Diphenylmethane
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-978-6
RTECSNumber DA4976500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only