missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. D22531G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22531G 1 g
D22535G 5 g
D225325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22531G Supplier TCI America Supplier No. D22531G

Chemical Identifiers

CAS 66003-76-7
Molecular Formula C13H10F3IO3S
Molecular Weight (g/mol) 430.18
MDL Number MFCD00191356
InChI Key SBQIJPBUMNWUKN-UHFFFAOYSA-M
Synonym diphenyliodonium trifluoromethanesulfonate, diphenyliodanium triflate, diphenyliodonium triflate, acmc-209nt6, diphenyliodonium trifluoromethansulfonat, diphenyliodanium; trifluoromethanesulfonate, diphenyliodonium trifluoromethane sulfonate, diphenyliodonium; trifluoromethanesulfonate, diphenyliodanium; tris fluoranyl methanesulfonate
PubChem CID 2737137
IUPAC Name diphenyliodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 1 g
Formula Weight 430.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Trifluoromethanesulfonate