accessibility menu, dialog, popup

UserName

CAS RN 66003-76-7

IUPAC Name: diphenyliodanium trifluoromethanesulfonate
Synonyms: diphenyliodanium triflate
Molecular Weight (g/mol): 430.18
Molecular Formula: C13H10F3IO3S
InChi Key: SBQIJPBUMNWUKN-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.[I+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)