missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. p-7127185
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22381G 1 g
D22385G 5 g
D223825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22381G Supplier TCI America Supplier No. D22381G
Only null left

Chemical Identifiers

CAS 58109-40-3
Molecular Formula C12H10I
Molecular Weight (g/mol) 281.12
MDL Number MFCD00061398
InChI Key OZLBDYMWFAHSOQ-UHFFFAOYSA-N
Synonym diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
PubChem CID 2737136
IUPAC Name diphenyliodanium
SMILES [I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 142°C
Color White
Quantity 1 g
Formula Weight 426.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Hexafluorophosphate