missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium chloride, 97%

CAS: 1483-72-3 | C12H10ClI | 316.57 g/mol

$354.10 - $354.10

Chemical Identifiers

CAS 1483-72-3
Molecular Formula C12H10ClI
Molecular Weight (g/mol) 316.57
MDL Number MFCD00011909
InChI Key RSJLWBUYLGJOBD-UHFFFAOYSA-M
Synonym diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
PubChem CID 73870
IUPAC Name diphenyliodanium;chloride
SMILES [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117330100
SDS View Documents
Thermo Scientific Chemicals
117330100
10 g Glass bottle
Each for $354.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1483-72-3
316.57
RSJLWBUYLGJOBD-UHFFFAOYSA-M
73870
[Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
C12H10ClI
MFCD00011909
diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
diphenyliodanium;chloride

Specifications

1483-72-3
Cream-Yellow to White
97%
C12H10ClI
MFCD00011909
01,341; 02,178
Solubility in water: soluble. Other solubilities: soluble in methanol
[Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
316.57
316.57
Powder
227°C to 228°C
Authentic
Glass bottle
(C6H5)2ICl
10 g
diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
RSJLWBUYLGJOBD-UHFFFAOYSA-M
diphenyliodanium;chloride
73870
97%
Diphenyliodonium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio

GHS Signal Word: Danger

EINECSNumber : 216-049-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only