Learn More
Diphenylcarbazide, 0.5% in Acetone, for Chromium, Certified, LabChem™
$105.39 - $105.39
Chemical Identifiers
| CAS | 140-22-7 |
|---|---|
| Molecular Formula | C13H14N4O |
| Molecular Weight (g/mol) | 242.282 |
| InChI Key | KSPIHGBHKVISFI-UHFFFAOYSA-N |
| Synonym | 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl |
| PubChem CID | 8789 |
| ChEBI | CHEBI:4641 |
| IUPAC Name | 1,3-dianilinourea |
| SMILES | C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
Chemical Identifiers
| 140-22-7 | |
| 242.282 | |
| 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
| CHEBI:4641 | |
| C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
| C13H14N4O | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| 8789 | |
| 1,3-dianilinourea |
Specifications
| Orange | |
| 99.5,0.5 | |
| C13H14N4O | |
| 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| 1,3-dianilinourea | |
| 8789 | |
| 242.28 | |
| Passes Test | |
| Passes Test | |
| 125 mL | |
| Diphenylcarbazide, 0.5% in Acetone, for Chromium |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature