missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine, 0.3% (w/v), Ricca Chemical
$177.47 - $177.47
Chemical Identifiers
| CAS | 6152-67-6 |
|---|---|
| Molecular Formula | C12H11NNaO3S |
| Molecular Weight (g/mol) | 272.27 |
| MDL Number | MFCD00036468 |
| InChI Key | VMVSDVIYIMNZBS-UHFFFAOYSA-N |
| Synonym | sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt |
| PubChem CID | 517002 |
| IUPAC Name | 4-(phenylamino)benzene-1-sulfonic acid sodium |
| SMILES | [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 6152-67-6 | |
| 272.27 | |
| VMVSDVIYIMNZBS-UHFFFAOYSA-N | |
| 517002 | |
| [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
| C12H11NNaO3S | |
| MFCD00036468 | |
| sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt | |
| 4-(phenylamino)benzene-1-sulfonic acid sodium |
Specifications
| Blue to Colorless | |
| 98.0,0.29 | |
| C12H11NNaO3S | |
| sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt | |
| [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 | |
| 272.27 | |
| 0.3% | |
| Amber Glass Bottle | |
| 120 mL | |
| 0.3% (w/v) |