missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylacetaldehyde 95.0+%, TCI America™
$262.83 - $1045.73
Chemical Identifiers
| CAS | 947-91-1 |
|---|---|
| Molecular Formula | C14H12O |
| Molecular Weight (g/mol) | 196.25 |
| MDL Number | MFCD00006972 |
| InChI Key | HLLGFGBLKOIZOM-UHFFFAOYSA-N |
| Synonym | diphenylacetaldehyde, diphenylketen, acetaldehyde, diphenyl, alpha-phenylbenzeneacetaldehyde, diphenylethanal, diphenyl-acetaldehyde, benzeneacetaldehyde, .alpha.-phenyl, diphenyl-acetaldehyd, benzeneacetaldehyde, ?-phenyl, diphenylacetoaldehyde |
| PubChem CID | 13696 |
| IUPAC Name | 2,2-diphenylacetaldehyde |
| SMILES | O=CC(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 947-91-1 | |
| 196.25 | |
| HLLGFGBLKOIZOM-UHFFFAOYSA-N | |
| 13696 | |
| O=CC(C1=CC=CC=C1)C1=CC=CC=C1 |
| C14H12O | |
| MFCD00006972 | |
| diphenylacetaldehyde, diphenylketen, acetaldehyde, diphenyl, alpha-phenylbenzeneacetaldehyde, diphenylethanal, diphenyl-acetaldehyde, benzeneacetaldehyde, .alpha.-phenyl, diphenyl-acetaldehyd, benzeneacetaldehyde, ?-phenyl, diphenylacetoaldehyde | |
| 2,2-diphenylacetaldehyde |
Specifications
| 947-91-1 | |
| C14H12O | |
| 5 g | |
| HLLGFGBLKOIZOM-UHFFFAOYSA-N | |
| 2,2-diphenylacetaldehyde | |
| 13696 | |
| ≥95.0% (GC) | |
| Diphenylacetaldehyde |
| 175°C | |
| MFCD00006972 | |
| diphenylacetaldehyde, diphenylketen, acetaldehyde, diphenyl, alpha-phenylbenzeneacetaldehyde, diphenylethanal, diphenyl-acetaldehyde, benzeneacetaldehyde, .alpha.-phenyl, diphenyl-acetaldehyd, benzeneacetaldehyde, ?-phenyl, diphenylacetoaldehyde | |
| O=CC(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 196.25 | |
| 196.25 | |
| Liquid |
Safety and Handling
RTECSNumber : AB2827500
TSCA : No