missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Sulfone 99.0+%, TCI America™

Catalog No. P023125G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P023125G 25 g
P0231500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. P023125G missing translation for 'mfr' TCI America missing translation for 'supplierNo' P023125G

Chemical Identifiers

CAS 127-63-9
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00007548
InChI Key KZTYYGOKRVBIMI-UHFFFAOYSA-N
Synonym diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
PubChem CID 31386
ChEBI CHEBI:78360
IUPAC Name (benzenesulfonyl)benzene
SMILES O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 127°C
Color White
Boiling Point 379°C
Quantity 25 g
Formula Weight 218.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Sulfone
EINECSNumber (3)-2158
RTECSNumber SX2400000
TSCA Yes