accessibility menu, dialog, popup

UserName

OSO

CAS RN 127-63-9

IUPAC Name: (benzenesulfonyl)benzene
Synonyms: diphenyl sulfone
Molecular Weight (g/mol): 218.27
Molecular Formula: C12H10O2S
InChi Key: KZTYYGOKRVBIMI-UHFFFAOYSA-N
SMILES: O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)