Learn More
Diphenyl sulfone, 99+%
CAS: 127-63-9 | C12H10O2S | 218.27 g/mol
Supplier: Thermo Scientific Chemicals L082320B
Description
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Diphenyl sulfone | |
| 123°C to 130°C | |
| 377°C to 379°C | |
| Odorless | |
| MFCD00007548 | |
| 1910573 | |
| diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone | |
| KZTYYGOKRVBIMI-UHFFFAOYSA-N | |
| benzenesulfonylbenzene | |
| 31386 | |
| 218.27 |
| 127-63-9 | |
| 1.36 | |
| 184°C (363°F) | |
| C12H10O2S | |
| 1000 g | |
| 14,3238 | |
| Insoluble in water. | |
| O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 218.27 | |
| CHEBI:78360 | |
| ≥99% |
Chemical Identifiers
| 127-63-9 | |
| 218.27 | |
| KZTYYGOKRVBIMI-UHFFFAOYSA-N | |
| 31386 | |
| benzenesulfonylbenzene |
| C12H10O2S | |
| MFCD00007548 | |
| diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone | |
| CHEBI:78360 | |
| O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 204-853-1
RTECSNumber : SX2400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only