missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfone, 99+%

Catalog No. AAL0823230
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0823230 250 g
AAL082320B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0823230 Supplier Thermo Scientific Chemicals Supplier No. L0823230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-63-9 | C12H10O2S | 218.27 g/mol

It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 127-63-9
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00007548
InChI Key KZTYYGOKRVBIMI-UHFFFAOYSA-N
Synonym diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
PubChem CID 31386
ChEBI CHEBI:78360
IUPAC Name benzenesulfonylbenzene
SMILES O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 123°C to 130°C
Density 1.36
Boiling Point 377°C to 379°C
Flash Point 184°C (363°F)
Odor Odorless
Quantity 250 g
Beilstein 1910573
Merck Index 14,3238
Solubility Information Insoluble in water.
Formula Weight 218.27
Percent Purity ≥99%
Chemical Name or Material Diphenyl sulfone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-853-1
RTECSNumber SX2400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only