missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfide, 98%

Catalog No. AAA1111414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1111414 25 g
AAA1111422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1111414 Supplier Thermo Scientific Chemicals Supplier No. A1111414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 139-66-2 | C12H10S | 186.27 g/mol

Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

Solubility
Soluble in ether, benzene, carbon disulphide.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 139-66-2
Molecular Formula C12H10S
Molecular Weight (g/mol) 186.27
MDL Number MFCD00003064
InChI Key LTYMSROWYAPPGB-UHFFFAOYSA-N
Synonym diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
PubChem CID 8766
ChEBI CHEBI:38959
IUPAC Name phenylsulfanylbenzene
SMILES S(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -40°C
Density 1.118
Boiling Point 295°C to 296°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.6325
Quantity 25 g
Beilstein 1907932
Merck Index 14,7314
Solubility Information Soluble in ether,benzene,carbon disulphide.
Formula Weight 186.28
Percent Purity 98%
Chemical Name or Material Diphenyl sulfide
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 205-371-4
RTECSNumber SX2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only