missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Oxalate 98.0+%, TCI America™

Catalog No. p-7134191
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O011125G 25 g
O0111100G 100 g
O0111500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O011125G Supplier TCI America Supplier No. O011125G
Only null left

Chemical Identifiers

CAS 3155-16-6
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00059682
InChI Key ULOZDEVJRTYKFE-UHFFFAOYSA-N
Synonym Oxalic Acid Diphenyl Ester
PubChem CID 18475
IUPAC Name diphenyl oxalate
SMILES C1=CC=C(C=C1)OC(=O)C(=O)OC2=CC=CC=C2

Specifications

Melting Point 136°C
Quantity 25 g
Formula Weight 242.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Oxalate
RTECSNumber RO2880000
TSCA No