accessibility menu, dialog, popup

UserName

OOOO

CAS RN 3155-16-6

IUPAC Name: diphenyl oxalate
Synonyms: diphenyl oxalate
Molecular Weight (g/mol): 242.23
Molecular Formula: C14H10O4
InChi Key: ULOZDEVJRTYKFE-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C(=O)OC1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results