missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Oxalate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Quantity | 25 g |
| Formula Weight | 242.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Diphenyl Oxalate |
Safety and Handling
| RTECSNumber | RO2880000 |
| TSCA | No |