missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate 95.0+%, TCI America™

Catalog No. D37091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37091G 1 g
D37095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37091G Supplier TCI America Supplier No. D37091G
Only null left

Chemical Identifiers

CAS 367508-01-8
Molecular Formula C16H18NO5P
Molecular Weight (g/mol) 335.30
MDL Number MFCD11042674
InChI Key CFSFGLDTIDFDMT-UHFFFAOYSA-N
Synonym (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diphenyl Ester
PubChem CID 12030864
IUPAC Name diphenyl {[methoxy(methyl)carbamoyl]methyl}phosphonate
SMILES CON(C)C(=O)CP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 335.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate