accessibility menu, dialog, popup

UserName

CH3NOPOOOOH3C

CAS RN 367508-01-8

IUPAC Name: diphenyl {[methoxy(methyl)carbamoyl]methyl}phosphonate
Synonyms: diphenyl [methoxy(methyl)carbamoyl]methylphosphonate
Molecular Weight (g/mol): 335.3
Molecular Formula: C16H18NO5P
InChi Key: CFSFGLDTIDFDMT-UHFFFAOYSA-N
SMILES: CON(C)C(=O)CP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results