missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenolic Acid 98.0+%, TCI America™
$74.56 - $870.51
Chemical Identifiers
| CAS | 126-00-1 |
|---|---|
| Molecular Formula | C17H18O4 |
| Molecular Weight (g/mol) | 286.327 |
| MDL Number | MFCD00002800 |
| InChI Key | VKOUCJUTMGHNOR-UHFFFAOYSA-N |
| Synonym | 4,4-Bis(4-hydroxyphenyl)valeric Acid |
| PubChem CID | 67174 |
| ChEBI | CHEBI:34721 |
| IUPAC Name | 4,4-bis(4-hydroxyphenyl)pentanoic acid |
| SMILES | CC(CCC(=O)O)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O |
Chemical Identifiers
| 126-00-1 | |
| 286.327 | |
| VKOUCJUTMGHNOR-UHFFFAOYSA-N | |
| 67174 | |
| 4,4-bis(4-hydroxyphenyl)pentanoic acid |
| C17H18O4 | |
| MFCD00002800 | |
| 4,4-Bis(4-hydroxyphenyl)valeric Acid | |
| CHEBI:34721 | |
| CC(CCC(=O)O)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O |
Specifications
| 126-00-1 | |
| White-Yellow | |
| MFCD00002800 | |
| 4,4-Bis(4-hydroxyphenyl)valeric Acid | |
| CC(CCC(=O)O)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O | |
| 286.327 | |
| CHEBI:34721 | |
| ≥98.0% (GC,T) | |
| Diphenolic Acid |
| 177°C | |
| C17H18O4 | |
| 25 g | |
| VKOUCJUTMGHNOR-UHFFFAOYSA-N | |
| 4,4-bis(4-hydroxyphenyl)pentanoic acid | |
| 67174 | |
| 286.33 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0264
TSCA : Yes