accessibility menu, dialog, popup

UserName

CH3HOOOHOH

CAS RN 126-00-1

IUPAC Name: 4,4-bis(4-hydroxyphenyl)pentanoic acid
Synonyms: diphenolic acid diphenolin acid
Molecular Weight (g/mol): 286.33
Molecular Formula: C17H18O4
InChi Key: VKOUCJUTMGHNOR-UHFFFAOYSA-N
SMILES: CC(CCC(O)=O)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results