missing translation for 'onlineSavingsMsg'
Learn More

Diphenolic Acid 98.0+%, TCI America™

Catalog No. D127425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D127425G 25 g
D1274500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D127425G Supplier TCI America Supplier No. D127425G

Chemical Identifiers

CAS 126-00-1
Molecular Formula C17H18O4
Molecular Weight (g/mol) 286.327
MDL Number MFCD00002800
InChI Key VKOUCJUTMGHNOR-UHFFFAOYSA-N
Synonym 4,4-Bis(4-hydroxyphenyl)valeric Acid
PubChem CID 67174
ChEBI CHEBI:34721
IUPAC Name 4,4-bis(4-hydroxyphenyl)pentanoic acid
SMILES CC(CCC(=O)O)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 25 g
Formula Weight 286.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenolic Acid
EINECSNumber (4)-0264
TSCA Yes