missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioctadecyl 3,3'-Thiodipropionate 90.0+%, TCI America™
$72.70 - $255.38
Chemical Identifiers
| CAS | 693-36-7 |
|---|---|
| Molecular Formula | C42H82O4S |
| Molecular Weight (g/mol) | 683.174 |
| MDL Number | MFCD00026684 |
| InChI Key | PWWSSIYVTQUJQQ-UHFFFAOYSA-N |
| PubChem CID | 12738 |
| IUPAC Name | octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate |
| SMILES | CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 693-36-7 | |
| 683.174 | |
| PWWSSIYVTQUJQQ-UHFFFAOYSA-N | |
| octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate |
| C42H82O4S | |
| MFCD00026684 | |
| 12738 | |
| CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC |
Specifications
| 693-36-7 | |
| White | |
| MFCD00026684 | |
| Distearyl 3,3′C-Thiodipropionate, 3,3′C-Thiodipropionic Acid Dioctadecyl Ester | |
| CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC | |
| 683.174 | |
| 683.17 | |
| Crystalline Powder |
| 65°C | |
| C42H82O4S | |
| 25 g | |
| PWWSSIYVTQUJQQ-UHFFFAOYSA-N | |
| octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate | |
| 12738 | |
| ≥90.0% (HPLC) | |
| Dioctadecyl 3,3′-Thiodipropionate |
Safety and Handling
EINECSNumber : (2)-0919&(2)-1399
RTECSNumber : UF8010000
TSCA : Yes