missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dinotefuran

CAS: 165252-70-0 | C7H14N4O3, C7H14N4O3 | 202.21 g/mol

$102.29 - $252.14

Chemical Identifiers

CAS 165252-70-0
Molecular Formula C7H14N4O3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC467090010
View Documents
Thermo Scientific Chemicals
467090010
1 g
Each for $252.14
Only null left
 
AC467092500
View Documents
Thermo Scientific Chemicals
467092500
250 mg
Each for $102.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

165252-70-0
C7H14N4O3

Specifications

165252-70-0
Solubility aq. soln.: 39.8 g/L (20°C). Other solubilities: soluble in ethanol, methanol and dimethyl sulfoxid, e
C7H14N4O3
YKBZOVFACRVRJN-UHFFFAOYNA-N
N''-methyl-N-nitro-N'-[(oxolan-3-yl)methyl]guanidine
Dinotefuran
1 g
106.0°C to 111.0°C
202.21
CN=C(NCC1CCOC1)N[N+]([O-])=O
202.21
Videos
Safety and Handling

Safety and Handling

GHS H Statement Very toxic to aquatic life with long lasting effects.

GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only