missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dinotefuran

Catalog No. AC467092500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467092500 250 mg
AC467090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467092500 Supplier Thermo Scientific Chemicals Supplier No. 467092500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 165252-70-0 | C7H14N4O3, C7H14N4O3 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165252-70-0
Molecular Formula C7H14N4O3

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: 39.8 g/L (20°C). Other solubilities: soluble in ethanol, methanol and dimethyl sulfoxid, e
Melting Point 106.0°C to 111.0°C
Formula Weight 202.21
Chemical Name or Material Dinotefuran
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only