missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphenylphosphine Borane 95.0+%, TCI America™
$180.97 - $1014.57
Chemical Identifiers
| CAS | 35512-87-9 |
|---|---|
| Molecular Formula | C8H11BP |
| Molecular Weight (g/mol) | 148.96 |
| MDL Number | MFCD28386099 |
| InChI Key | QZAXONBUUURRQX-UHFFFAOYSA-N |
| Synonym | Borane - Dimethylphenylphosphine Complex |
| PubChem CID | 11412474 |
| IUPAC Name | dimethyl(phenyl)phosphane boron |
| SMILES | [B].CP(C)C1=CC=CC=C1 |
Chemical Identifiers
| 35512-87-9 | |
| 148.96 | |
| QZAXONBUUURRQX-UHFFFAOYSA-N | |
| 11412474 | |
| [B].CP(C)C1=CC=CC=C1 |
| C8H11BP | |
| MFCD28386099 | |
| Borane - Dimethylphenylphosphine Complex | |
| dimethyl(phenyl)phosphane boron |
Specifications
| 35512-87-9 | |
| 78°C | |
| MFCD28386099 | |
| Borane - Dimethylphenylphosphine Complex | |
| [B].CP(C)C1=CC=CC=C1 | |
| 148.96 | |
| 151.98 | |
| Liquid |
| Yellow | |
| C8H11BP | |
| 1 g | |
| QZAXONBUUURRQX-UHFFFAOYSA-N | |
| dimethyl(phenyl)phosphane boron | |
| 11412474 | |
| ≥95.0% (HPLC) | |
| Dimethylphenylphosphine Borane |
Safety and Handling
TSCA : No