missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylphosphine Borane 95.0+%, TCI America™

Catalog No. d42041g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42041G 1 g
D42045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42041G Supplier TCI America Supplier No. D42041G

Chemical Identifiers

CAS 35512-87-9
Molecular Formula C8H11BP
Molecular Weight (g/mol) 148.96
MDL Number MFCD28386099
InChI Key QZAXONBUUURRQX-UHFFFAOYSA-N
Synonym Borane - Dimethylphenylphosphine Complex
PubChem CID 11412474
IUPAC Name dimethyl(phenyl)phosphane boron
SMILES [B].CP(C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 1 g
Formula Weight 151.98
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Dimethylphenylphosphine Borane