missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylisopropylchlorosilane 90.0+%, TCI America™
Supplier: TCI America D15945ML
Specifications
| Dimethylisopropylchlorosilane [Dimethylisopropylsilylating Agent] | |
| Yellow | |
| C5H13ClSi | |
| 5 mL | |
| isopropyldimethylchlorosilane, dimethylisopropylchlorosilane, chloro isopropyl dimethylsilane, chloro dimethyl isopropylsilane, chlorodimethylisopropylsilane, silane,chlorodimethyl 1-methylethyl, silane, chlorodimethyl 1-methylethyl, dmipscl, acmc-1bn7i, chlorodimethyl isopropylsilane | |
| CC(C)[Si](C)(C)Cl | |
| 136.69 | |
| 136.69 | |
| Liquid |
| 3634-56-8 | |
| 112°C | |
| MFCD00009917 | |
| 2985 | |
| YCXVDEMHEKQQCI-UHFFFAOYSA-N | |
| chlorodimethyl(propan-2-yl)silane | |
| 5102883 | |
| ≥90.0% (GC) |
Chemical Identifiers
| 3634-56-8 | |
| 136.69 | |
| YCXVDEMHEKQQCI-UHFFFAOYSA-N | |
| 5102883 | |
| CC(C)[Si](C)(C)Cl |
| C5H13ClSi | |
| MFCD00009917 | |
| isopropyldimethylchlorosilane, dimethylisopropylchlorosilane, chloro isopropyl dimethylsilane, chloro dimethyl isopropylsilane, chlorodimethylisopropylsilane, silane,chlorodimethyl 1-methylethyl, silane, chlorodimethyl 1-methylethyl, dmipscl, acmc-1bn7i, chlorodimethyl isopropylsilane | |
| chlorodimethyl(propan-2-yl)silane |
Safety and Handling
EINECSNumber : (2)-2041
TSCA : No