missing translation for 'onlineSavingsMsg'
Learn More

Dimethylisopropylchlorosilane 90.0+%, TCI America™

Catalog No. p-7126813
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15945ML 5 mL
D159425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15945ML Supplier TCI America Supplier No. D15945ML
Only null left

Chemical Identifiers

CAS 3634-56-8
Molecular Formula C5H13ClSi
Molecular Weight (g/mol) 136.69
MDL Number MFCD00009917
InChI Key YCXVDEMHEKQQCI-UHFFFAOYSA-N
Synonym isopropyldimethylchlorosilane, dimethylisopropylchlorosilane, chloro isopropyl dimethylsilane, chloro dimethyl isopropylsilane, chlorodimethylisopropylsilane, silane,chlorodimethyl 1-methylethyl, silane, chlorodimethyl 1-methylethyl, dmipscl, acmc-1bn7i, chlorodimethyl isopropylsilane
PubChem CID 5102883
IUPAC Name chlorodimethyl(propan-2-yl)silane
SMILES CC(C)[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 mL
UN Number 2985
Formula Weight 136.69
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethylisopropylchlorosilane [Dimethylisopropylsilylating Agent]
EINECSNumber (2)-2041
TSCA No