accessibility menu, dialog, popup

UserName

H3CCH3SiCH3CH3Cl

CAS RN 3634-56-8

IUPAC Name: chlorodimethyl(propan-2-yl)silane
Synonyms: chloro(isopropyl)dimethylsilane
Molecular Weight (g/mol): 136.69
Molecular Formula: C5H13ClSi
InChi Key: YCXVDEMHEKQQCI-UHFFFAOYSA-N
SMILES: CC(C)[Si](C)(C)Cl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)