missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime RS, 1% (w/v) Alcoholic Solution, Ricca Chemical

Catalog No. p-3251612
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25704 120 mL Natural Poly Bottle
257016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25704 Supplier Ricca Chemical Company Supplier No. RI25704
Only null left

For Nickel Determination

Chemical Identifiers

CAS 95-45-4 , 64-17-5 , 67-56-1
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 135399895
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Color Colorless
CAS Min % 1.21, 97.0
CAS Max % 1.26, 100.0
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Alcoholic Solution
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Dimethylglyoxime, Reagent Solution
Show More Show Less